Iron(III) nitrate nonahydrate 99.999%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7782-61-8
:
Formula:Fe(NO₃)₃·9H₂O
MDL Number:MFCD00149708
:404 g/mol
Boiling Pt:125 °C
Melting Pt:47.2 °C
Density:1.684 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1466
ADR:5.1,III
:GHS02
:GHS07
Frequently Bought Together