Calcium chloride 77%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10043-52-4
:
Formula:CaCl₂
MDL Number:MFCD00010903
Boiling Pt:1670 °C
Melting Pt:772 °C
Density:2.15 g/cm³ (25 °C)
:110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
:GHS07
Frequently Bought Together