Order Entry
Austria
ContactUsLinkComponent
Ammonium thiocyanate 98+%
Irritant
Ammonium thiocyanate 98+%
Catalog # APOSIN1093-1KG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ammonium thiocyanate 98+%
Catalog # APOSIN1093-1KG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :76.12 g/mol
Melting Pt:152…154 °C
Density:1.3 g/cm³ (25 °C)
EINECS:217-175-6
 :GHS07
Storage Temperature:Ambient
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428