Order Entry
Austria
Orders LinkContactUsLinkComponent
Vancomycin hydrochloride

Vancomycin hydrochloride

Irritant
Supplier:   Apollo Scientific
Vancomycin hydrochloride
Supplier:  Apollo Scientific
Certificates
Vancomycin hydrochloride

3 Options Available Below

Product Details & Documents

Vancomycin is a glycopeptide antibiotic. It exerts its action by inhibiting the formation of the peptidoglycan polymers of the bacterial cell wall. Often used incombination with cefotaxim or carbenicillin.

  • Potency of 1099 mcg/mg
  • Cell culture tested.
Synonyms:  (3S,6R,7R,22R,23S,26S,36R,38aR)-3-(2-Amino-2-oxoethyl)-44-[[2-O-(3-amino-2,3,6-trideoxy-3-C-methyl-α-L-lyxo-hexopyranosyl)-β-D-glucopyranosyl]oxy]-10,19-dichloro-2,3,4,5,6,7,23,24,25,26,36,37,38,38a-tetradecahydro-7,22,28,30,32-pentahydroxy-6-[[(2R)-4-methyl-2-(methylamino)-1-oxopentyl]amino]-2,5,24,38,39-pentaoxo-22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine-26-carboxylic acid xhydrochloride, Vancomycin supplement
  • CAS Number:1404-93-9
  • Formula:C₆₆H₇₅Cl₂N₉O₂₄·HCl
  • MDL Number:MFCD03613611
  • Molecular Weight:1485.71 g/mol
  • Storage Temperature:Refrigerator
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 29,00 €
    29,00 €
    1 * 1 g
       29,00 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 110,00 €
    110,00 €
    1 * 5 g
       110,00 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 261,00 €
    261,00 €
    1 * 25 g
       261,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.