Order Entry
Austria
ContactUsLinkComponent
Tween® 20 (Polysorbate)
Tween® 20 (Polysorbate)
Catalog # APOSBIT0803-1L
Supplier:  Apollo Scientific
CAS Number:  
Tween® 20 (Polysorbate)
Catalog # APOSBIT0803-1L
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :1227.53 g/mol
Boiling Pt:>100 °C
Melting Pt:–14 °C
Density:1.095 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:500-018-3
CAS Number:9005-64-5
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986