Order Entry
Austria
ContactUsLinkComponent
Ponceau S
Irritant
Ponceau S
Catalog # APOSBIP1405-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ponceau S
Catalog # APOSBIP1405-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :760.56 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:6226-79-5
Formula:C₂₂H₁₂N₄Na₄O₁₃S₄
MDL Number:MFCD00003892