Toluene-α-sulphonyl fluoride 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:329-98-6
:
Formula:C₇H₇FO₂S
MDL Number:MFCD00007424
:174.2 g/mol
Melting Pt:92…94 °C
Storage Temperature:Ambient
EINECS:206-350-2
UN:2928
ADR:6.1+8,II
:GHS05
:GHS06
Frequently Bought Together