Order Entry
Austria
Orders LinkContactUsLinkComponent
β-Cyclodextrine

β-Cyclodextrine

Irritant
Supplier:   Apollo Scientific
β-Cyclodextrine
Supplier:  Apollo Scientific
Certificates
β-Cyclodextrine

3 Options Available Below

Product Details & Documents
Synonyms:  Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex
  • CAS Number:7585-39-9
  • Formula:C₄₂H₇₀O₃₅
  • MDL Number:MFCD00078139
  • Molecular Weight:1134.98 g/mol
  • Melting Pt:290…300 °C
  • Density:1.44 g/cm³ (25 °C)
  • Storage Temperature:Ambient

Documents

Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.