Order Entry
Austria
ContactUsLinkComponent
Hygromycin B in PBS
Toxic
Corrosive
Health
Hygromycin B in PBS
Catalog # APOSBIH0125-20ML
Supplier:  Apollo Scientific
CAS Number:  
Toxic
Corrosive
Health
Hygromycin B in PBS
Catalog # APOSBIH0125-20ML
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    20 mL
  • Seq:
    2305

Specifications

About this item

 :
 :527.5 g/mol
Storage Temperature:Refrigerator
UN:3462
ADR:6.1,I
 :GHS06
 :GHS05
 :GHS08
CAS Number:31282-04-9
Formula:C₂₀H₃₇N₃O₁₃
MDL Number:MFCD06795479