Order Entry
Austria
Orders LinkContactUsLinkComponent
D-(+)-Galactose

D-(+)-Galactose

Supplier:   Apollo Scientific
D-(+)-Galactose
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00151230
  • Molecular Weight:180.16 g/mol
  • Melting Pt:168…170 °C
  • Density:1.5 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:200-416-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 50,70 €
    50,70 €
    1 * 500 g
       50,70 €

  • Bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 86,90 €
    86,90 €
    1 * 1 kg
       86,90 €

  • Bottle
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 398,00 €
    398,00 €
    1 * 5 kg
       398,00 €