

Evans blue
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:314-13-6
:
Formula:C₃₄H₂₈N₆O₁₄S₄
MDL Number:MFCD00004021
ADR:4.1,III
:960.79 g/mol
EINECS:206-242-5
UN:1325
:GHS02
:GHS08
Storage Temperature:Ambient
Frequently Bought Together