Order Entry
Austria
ContactUsLinkComponent
2'-Deoxyadenosine-5'-triphosphate disodium salt (dATP disodium salt) 97+%
Irritant
2'-Deoxyadenosine-5'-triphosphate disodium salt (dATP disodium salt) 97+%
Catalog # APOSBID2033-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2'-Deoxyadenosine-5'-triphosphate disodium salt (dATP disodium salt) 97+%
Catalog # APOSBID2033-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :535.14 g/mol
EINECS:277-809-2
 :GHS07
Storage Temperature:Freezer
CAS Number:74299-50-6
Formula:C₁₀H₁₄N₅Na₂O₁₂P₃
MDL Number:MFCD00080335