0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
DSP (Dithiobis(succinimidyl propionate))
DSP (Dithiobis(succinimidyl propionate))
DSP (Dithiobis(succinimidyl propionate))
APOSBICL102-1G
:
Apollo Scientific
:
DSP (Dithiobis(succinimidyl propionate))
APOSBICL102-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
404.41 g/mol
:
128…133 °C
:
260-931-5
:
GHS07
:
Refrigerator
:
57757-57-0
:
C₁₄H₁₆N₂O₈S₂
:
MFCD00042045