
3'-Deoxyadenosine (cordycepine)
Catalog # APOSBIC1025-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:251.24 g/mol
Melting Pt:225 °C
Storage Temperature:Freezer
UN:3462
ADR:6.1,III
:GHS06
CAS Number:73-03-0
Formula:C₁₀H₁₃N₅O₃
MDL Number:MFCD00037998