
L(+)-Cysteine hydrochloride monohydrate 99+%
Frequently Bought Together
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
L(+)-Cysteine hydrochloride monohydrate, TECHNICAL
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Silver nitrate 99.9+%
: Apollo Scientific
Cetrimonium bromide ≥98%
: Thermo Fisher Scientific
VWR® Cellulose Stoppers
: VWR Collection