Tetradecyltrimethylammonium bromide BP
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1119-97-7
:8044-71-1
Formula:C₁₇H₃₈BrN
MDL Number:MFCD00011770
:336.394 g/mol
Melting Pt:245…250 °C
Storage Temperature:Ambient
UN:3263
ADR:8,II
Frequently Bought Together