
7-Amino-4-methylcoumarin
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:26093-31-2
:
Formula:C₁₀H₉NO₂
MDL Number:MFCD00006868
:175.19 g/mol
Melting Pt:223…226 °C
EINECS:247-454-8
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together