Order Entry
Austria
Orders LinkContactUsLinkComponent
Formaldehyde

Formaldehyde

Flammable
Corrosive
Toxic
Health
New Product
Supplier:   Apollo Scientific
Formaldehyde
Supplier:  Apollo Scientific
New Product
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Boiling Pt:93…96 °C
  • Density:1.08…1.1 g/cm³ (25 °C)
  • Flash Pt:56 °C
  • Storage Temperature:Ambient
  • UN:2209
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 86,90 €
    86,90 €
    1 * 25 mL
       86,90 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 141,00 €
    141,00 €
    1 * 500 mL
       141,00 €

    Specifications:
    New Product
    New Product