Order Entry
Austria
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid ≥99.5%

L(+)-Ascorbic acid ≥99.5%

Corrosive
Irritant
Supplier:   Apollo Scientific
L(+)-Ascorbic acid ≥99.5%
Supplier:  Apollo Scientific
Certificates
L(+)-Ascorbic acid ≥99.5%

2 Options Available Below

Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Melting Pt:190…194 °C (dec.)
  • Density:1.65 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:200-066-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 26,10 €
    26,10 €
    1 * 500 g
       26,10 €

  • Bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 46,40 €
    46,40 €
    1 * 1 kg
       46,40 €