
4-Aminobenzoic acid 99%
:150-13-0
:
:C₇H₇NO₂
:MFCD00007894
:137.14 g/mol
:186…189 °C
:3.5 g/cm³ (25 °C)
:GHS07
:Ambient
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Transfer pipettes
: RATIOLAB
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR®, Aluminium foil in dispensers
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection