Order Entry
Austria
ContactUsLinkComponent
Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP) 99%
Irritant
Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP) 99%
Catalog # APOSBI3332-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Lithium phenyl(2,4,6-trimethylbenzoyl)phosphinate (LAP) 99%
Catalog # APOSBI3332-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :294.21 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:85073-19-4
Formula:C₁₆H₁₆LiO₃P
MDL Number:MFCD29472537