Orders LinkContactUsLinkComponent
qScript® ultra flex kits
Supplier:   Quantabio
qScript® ultra flex kits
Supplier:  Quantabio
Certificates
qScript® ultra flex kits
qScript® ultra flex kits
2 Options Available Below
Product Details & Documents
Faster, longer, tougher next gen cDNA synthesis with flexible priming.
  • Enhanced thermostability – Engineered for RT activity up to 65 °C, overcoming challenging secondary structures
  • Superior speed and ease of use – 10 minute reaction time with fewer pipetting steps
  • Maximum yield and sensitivity – Improved cDNA yield with limiting or compromised samples
  • Ultimate inhibitor resistance – Overcome a wide array of PCR inhibitors (salt, heparin, hematin, etc.)
  • Longer transcripts – Ability to reverse transcribe single targets up to 20 kb
qScript Ultra Flex Kit is easy-to-use and highly efficient for the synthesis of first-strand cDNA with flexible priming methods including Oligo-dT, Random Hexamer and Gene Specific Priming to reverse transcribe RNA to cDNA. A key component is a novel, state-of-the-art, RNase H deficient reverse transcriptase that was engineered for improved thermostability, velocity, processivity, and resistance to many common reaction inhibitors. qScript Ultra Flex Kit contains all required components for first-strand cDNA synthesis except RNA template and gene specific primers. The cDNA product is directly compatible with downstream 2-step RT-qPCR or RT-PCR procedures.
qScript Ultra Flex Kit has an optimal reaction temperature of 55°C and a 10-min reaction time, significantly reducing time to result while working through challenging secondary RNA structures. The lower Cq values for the engineered qScript Ultra reverse transcriptase indicate more efficient cDNA synthesis at high temperatures, critical for progressing through high GC regions and regions of RNA secondary structure.
Product Family Options
Product Information
  • Tests per kit
    Pk
    Availability
    Price
  • 733-2898
    qScript® Ultra Flex kit
    qScript® Ultra Flex kit
    25 Reactions
    25 Tests
    Log in for availability
    Log in for availability
    - 86,20 €
    86,20 €
       86,20 €
  • 733-2899
    qScript® Ultra Flex kit
    qScript® Ultra Flex kit
    100 Reactions
    100 Tests
    Log in for availability
    Log in for availability
    - 307,00 €
    307,00 €
       307,00 €