Order Entry
Austria
ContactUsLinkComponent
Human recombinant CD36 (from HEK293)
Human recombinant CD36 (from HEK293)
Catalog # ACRBCD6-H5221500UG
Supplier:  ACRO Biosystems
CAS Number:  
Human recombinant CD36 (from HEK293)
Catalog # ACRBCD6-H5221500UG
Supplier:  ACRO Biosystems
CAS Number:  

Specifications

  • Pack type:
    500 µg
  • Pk:
    500 µG
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.; Human CD36, His Tag immobilised on CM5 Chip can bind Human Thrombospondin-2, His Tag with an affinity constant of 21 nM as determined in a SPR assay (Biacore 8K) (Routinely tested)
  • Gene ID:
    P16671
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    GP4|GP3B|FAT|GPIV|gpIV|SCARB3|CHDS7|glycoprotein IIIb|CD36|glycoprotein IV|BDPLT10|gpIIIb|PASIV|Platelet Glycoprotein 4
  • Protein/peptide name:
    CD36
  • Purity:
    >95%
  • Molecular weight:
    48,5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

About this item