Order Entry
Austria
ContactUsLinkComponent
Human Recombinant CD229 (from HEK293 cells), Biotin
Human Recombinant CD229 (from HEK293 cells), Biotin
Catalog # ACRBCD9-H82E6-25UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  
Human Recombinant CD229 (from HEK293 cells), Biotin
Catalog # ACRBCD9-H82E6-25UG
Supplier:  ACROBIOSYSTEMS
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His, Avitag™
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Gene ID:
    Q9HBG7-1
  • Protein synonyms:
    LY9|hly9|Ly-9|SLAMF3|Ly9
  • Protein/peptide name:
    CD229
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    48,5 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
  • Shipping temperature:
    RT

Specifications

About this item

Biotinylated Human CD229 / SLAMF3 protein, His,Avitag™.