Human recombinant BAFF (from HEK293 cells)
Catalog # ACRBBAF-H52D4-1MG
Supplier: ACROBIOSYSTEMS
CAS Number:
Specifications
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Seq:02
- Tag sequence:active trimer (MALS verified)
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Human BAFFR Protein, Fc Tag (Cat. No. BAR-H5257) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human BAFF Protein, His,Flag Tag (Cat. No. BAF-H52D4) with an affinity constant of 5,54 nM as determined in a SPR assay (Biacore T200) (Routinely tested). Human BAFF, His,Flag Tag, active trimer (Cat. No. BAF-H52D4) stimulates proliferation of mouse spleen cells. The EC50 for this effect is 1,97-2,90 ng/ml (Routinely tested).
- Protein synonyms:CD257|ZTNF4|THANK|TALL-1|TNFSF20|DTL|TNFSF13B|TALL1|BLYS|BAFF
- Protein/peptide name:BAFF
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:52,5 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 25 mM MES, 150 mM NaCl, pH 6,0. Normally Trehalose is added as protectant before Lyophilisation.
- Shipping temperature:RT
Specifications
About this item
Human BAFF / TNFSF13B / CD257 protein, His,Flag Tag, active trimer (MALS verified).