Human Recombinant 4-1BB (from HEK293 cells), Biotin
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Biotinylated Human 4-1BB / TNFRSF9 protein, His,Avitag™(MALS verified).
Specifications
- Conjugation:Biotin
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:His, Avitag™
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Biotinylated Human 4-1BB Protein, His,Avitag™;;;;;;;
- Gene ID:Q07011-1
- Protein synonyms:CD137|TNFRSF9|ILA|CDw137|4-1BB
- Protein/peptide name:4-1BB
- Purity:>90% as determined by SDS-PAGE.
- Molecular weight:20,9 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
- Shipping temperature:RT
Specifications
Frequently Bought Together