Order Entry
Austria
ContactUsLinkComponent
Rabbit recombinant Serum Albumin (from HEK293 cells)
Rabbit recombinant Serum Albumin (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Rabbit recombinant Serum Albumin (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Rabbit
  • Tag sequence:
    His Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Loaded Biotinylated Human FCGRT and B2M Heterodimer protein, Avitag,His Tag and Strep II Tag (SPR and BLI verified)(Cat. No. FCM-H82W4) on SA Biosensor, can bind Rabbit Serum Albumin, His Tag(Cat. No. RSA-R52H6) with an affinity constant of 0,331 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    P49065-1
  • Protein synonyms:
    Serum albumin|Alb|ALB
  • Protein/peptide name:
    Serum Albumin
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    67,9 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together