Order Entry
Austria
ContactUsLinkComponent
Human recombinant integrin alpha M beta 2 (from HEK293 cells)
Human recombinant integrin alpha M beta 2 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant integrin alpha M beta 2 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag&Tag Free
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Gene ID:
    P11215-2 (ITGAM) & P05107-1 (ITGB2)
  • Protein synonyms:
    ITGAM&ITGB2|Integrin alpha M beta 2
  • Protein/peptide name:
    Integrin alpha M beta 2
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    127,2 kDa (ITGAM) and 80,2 kDa (ITGB2)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH7,5. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together