Order Entry
Austria
ContactUsLinkComponent
Human recombinant LAG-3 (from HEK293)
Human recombinant LAG-3 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant LAG-3 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised human LAG-3, mouse IgG2a Fc Tag (Cat. No. ACRBLA3-H52Aa) at 2 μg/ml (100 μl/well) can bind anti-human LAG3 MAb, human IgG1 with a linear range of 1 - 10 ng/ml (QC tested). Measured by its binding ability in a functional ELISA. Immobilised human LAG-3, mouse IgG2a Fc Tag (Cat. No. ACRBLA3-H52Aa) at 10 μg/ml (100 μl/well) can bind cynomolgus / rhesus macaque FGL1, Fc Tag (Cat. No. ACRBFG1-C5269) with a linear range of 0,01 - 0,156 μg/ml (routinely tested). Flow cytometry assay shows that Human LAG3, Mouse IgG2a Fc Tag (Cat. No. ACRBLA3-H52Aa) can bind to daudi cell surface. The concentration of LAG3 used is 0,3 μg/ml (routinely tested). FACS analysis shows that the binding Human LAG3, mouse IgG2a Fc Tag (Cat. No. ACRBLA3-H52Aa) to daudi cells was inhibited by increasing concentration of neutralising Anti-LAG3 antibody. The concentration of LAG3 used is 1 μg/ml. The IC50 is 0,165 μg/ml (routinely tested).
  • Gene ID:
    P18627-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    FDC|CD223|LAG3
  • Protein/peptide name:
    LAG-3
  • Purity:
    >95%
  • Molecular weight:
    73,3 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together