Human recombinant integrin isoform alpha-7X2B beta 1 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Gene ID:Q13683-7 (ITGA7X2B) and P05556-1 (ITGB1)
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:ITGA7X2B and ITGB1|ITGA7X2BB1|Integrin isoform alpha-7X2B beta 1
- Protein/peptide name:Integrin isoform alpha-7X2B beta 1
- Purity:>90%
- Molecular weight:116,5 kDa (ITGA7X2B) and 83,7 kDa (ITGB1)
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together