Order Entry
Austria
ContactUsLinkComponent
Human recombinant integrin alpha V beta 8 (from HEK293)
Human recombinant integrin alpha V beta 8 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant integrin alpha V beta 8 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 8,0. Normally trehalose is added as protectant before lyophilisation.; Measured by its binding ability in a functional ELISA. Immobilised human TGFB1, His tag (Cat. No. ACRBTG1-H524x) at 5 μg/ml (100 μl/well) can bind biotinylated human ITGAV and ITGB8 heterodimer protein, His, Avitag and tag free (Cat. No. ACRBIT8-H82W5) with a linear range of 5 - 156 ng/ml (QC tested).
  • Gene ID:
    P06756-1(V783I) (ITGAV) and P26012-1 (ITGB8)
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    Integrin alpha V beta 8|ITGAV and ITGB8
  • Protein/peptide name:
    Integrin alpha V beta 8
  • Purity:
    >90%
  • Molecular weight:
    114,7 kDa (ITGAV) and 76,5 kDa (ITGB8)
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 8,0. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together