Order Entry
Austria
ContactUsLinkComponent
Human recombinant NRG1 Beta 1 (from HEK293 cells)
Human recombinant NRG1 Beta 1 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant NRG1 Beta 1 (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    Fc Tag
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Gene ID:
    Q02297-6
  • Protein synonyms:
    Neuregulin1 beta 1|NDF|SMDF|ARIA|NRG1 Beta 1|NRG1|HRG1 beta 1|GGF|Neuregulin-1 beta 1|HRGA|HGL
  • Protein/peptide name:
    NRG1 Beta 1
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    53,2 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together