Order Entry
Austria
ContactUsLinkComponent
 
HCoV-HKU1(isolate N5) recombinant S1 protein (from HEK293 cells)
 
HCoV-HKU1(isolate N5) recombinant S1 protein (from HEK293 cells)

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    HCoV-HKU1(isolate N5)
  • Tag sequence:
    His Tag
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Gene ID:
    Q0ZME7-1
  • Protein synonyms:
    Spike glycoprotein Subunit1|Spike protein S1, COVID-19|S glycoprotein Subunit1|Spike, S1 protein
  • Protein/peptide name:
    S1 protein
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    85,7 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

 

Frequently Bought Together