Order Entry
Austria
ContactUsLinkComponent
Cynomolgus monkey recombinant IL-17A (from HEK293 cells)
Cynomolgus monkey recombinant IL-17A (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS
Certificates
Cynomolgus monkey recombinant IL-17A (from HEK293 cells)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Tag sequence:
    His Tag
  • Storage conditions:
    −20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method
  • Gene ID:
    G7P4U9-1
  • Protein synonyms:
    IL-17|IL-17A|CTLA-8|Interleukin-17A
  • Protein/peptide name:
    IL-17A
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular weight:
    17,0 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together