Human recombinant HVEM (from HEK293 cells), Fc tag, Acro Biosystems
- Conjugation:Unconjugated
- Protein function:Immune Checkpoint Proteins
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Tag sequence:Fc Tag
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human LIGHT, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. LIT-H5256) at 2 μg/ml (100 μl/well) can bind Human HVEM, Fc Tag (Cat. No. HVM-H5258) with a linear range of 0,3 - 20 ng/ml (QC tested).; Measured by its binding ability in a functional ELISA. Immobilised Human HVEM, Fc Tag (Cat. No. HVM-H5258) at 5 μg/ml (100 μl/well) can bind Biotinylated Human BTLA, Fc Tag (Cat. No. BTA-H82F8) with a linear range of 0,156 - 5 μg/ml (Routinely tested).
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:HVEA|HVEM|CD270|ATAR|TNFRSF14|TR2|LIGHTR
- UniProtKB:Q92956
- Protein/peptide name:HVEM
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:45,4 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
- Shipping temperature:Powder,RT
Frequently Bought Together