Human recombinant G-CSF R (from HEK293), Acro Biosystems
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised ActiveMax® Human G-CSF, Tag Free (Cat. No. ACRBGCF-H5214) at 2 μg/ml (100 μl/well) can bind Biotinylated Human G-CSF R, Avitag,His Tag (Cat. No. ACRBGCR-H82E4) with a linear range of 0,156 - 1,25 μg/ml (QC tested).
- Gene ID:Q99062
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CSF3R|GCSFR|CD114
- Protein/peptide name:G-CSF R
- Purity:>95%
- Molecular weight:69,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together