Human recombinant CEACAM-5 (from HEK293), Acro Biosystems
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Gene ID:P06731
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:Meconium antigen 100|CEA|CD66e|CEACAM-5
- Protein/peptide name:CEACAM-5
- Purity:>95%
- Molecular weight:72,2 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together