Order Entry
Austria
ContactUsLinkComponent
Human recombinant CD40 ligand (from HEK293), Acro Biosystems
Human recombinant CD40 ligand (from HEK293), Acro Biosystems
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant CD40 ligand (from HEK293), Acro Biosystems
Supplier:  ACROBIOSYSTEMS

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human CD40, His Tag (Cat. No. ACRBCD0-H5228) at 2 μg/ml (100 μl/well) can bind Biotinylated Human CD40 Ligand, Avitag,Fc Tag (Cat. No. ACRBCDL-H82F1) with a linear range of 1 - 31 ng/ml (QC tested).
  • Gene ID:
    P29965-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    TRAP|CD40L|IMD3|gp39|CD154|CD40LG|IGM|T-BAM|TNFSF5|HIGM1
  • Protein/peptide name:
    CD40 Ligand
  • Purity:
    >95%
  • Molecular weight:
    45,0 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together