Order Entry
Austria
ContactUsLinkComponent
Human recombinant CD40 (from HEK293), Acro Biosystems
Human recombinant CD40 (from HEK293), Acro Biosystems
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant CD40 (from HEK293), Acro Biosystems
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 0,1 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation. Measured by its binding ability in a functional ELISA. Immobilised Human CD40 Ligand, Fc Tag (Cat. No. ACRBCDL-H5267) at 5 μg/ml (100 μl/well) can bind Human CD40, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. ACRBCD0-H525a) with a linear range of 20 - 156 ng/ml (QC tested). Loaded Human CD40, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. ACRBCD0-H525a) on Protein A Biosensor, can bind Human CD40 Ligand, His,Flag Tag (active trimer) (MALS verified) (Cat. No. ACRBCDL-H52Db) with an affinity constant of 0,911 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested). FACS analysis shows that Human CD40, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. ACRBCD0-H525a) can bind to 293T cells overexpressing human CD40L. The concentration of Human CD40 is 0,03 μg/ml (Routinely tested).
  • Gene ID:
    P25942-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    p50|CD40|MGC9013|Bp50|TNFRSF5|CDW40
  • Protein/peptide name:
    CD40
  • Purity:
    >95%
  • Molecular weight:
    46,1 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in 50 mM Tris, 100 mM Glycine, 150 mM NaCl, pH 7,5. Normally trehalose is added as protectant before lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together