Human recombinant PD-1 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 0,1 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4, Normally trehalose is added as protectant before Lyophilisation,; Measured by its binding ability in a functional ELISA, Immobilised Human PD-L1, Fc Tag (HPLC-verified) (Cat, No, PD1-H5258) at 10 μg/ml (100 μl/well) can bind Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat, No, PD1-H5255) with a linear range of 2-40 ng/ml (QC tested),; Measured by its binding ability in a functional ELISA, Immobilised Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat, No, PD1-H5255) at 2 μg/ml (100 μl/well) can bind Human PD-L2, Fc Tag (Cat, No, PD2-H5251) with a linear range of 0,2-4 ng/ml (Routinely tested),; Measured by its binding ability in a functional ELISA, Immobilised Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat, No, PD1-H5255) at 2 μg/ml (100 μl/well) can bind Nivolumab with a linear range of 0,1-2 ng/ml (Routinely tested),; Measured by its binding ability in a functional ELISA, Immobilised Human PD-L1, His Tag (HPLC-verified) (Cat, No, PD1-H5229) at series of concentration on MonRabTM Anti-His Tag (C-term) Antibody precoated (0,1 μg/well) plate, can bind Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat, No, PD1-H5255) at 5 μg/ml (100 μl/well) with a linear range of 10-78 ng/ml (Routinely tested),; Loaded Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat, No, PD1-H5255) on Protein A Biosensor, can bind Human PD-L1, His Tag (HPLC verified) (Cat, No, PD1-H5229) with an affinity constant of 2,6 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested),; Loaded Human PD-1, Mouse IgG2a Fc Tag (HPLC-verified) (Cat, No, PD1-H5255) on Protein A Biosensor, can bind Human PD-L2, His Tag (SPR verified) (Cat, No, PD2-H5220) with an affinity constant of 0,43 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q15116-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:CD279|PDCD1|PD1|SLEB2
- Protein/peptide name:PD-1
- Purity:>95%
- Molecular weight:42,5 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4, Normally trehalose is added as protectant before Lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together