Order Entry
Austria
ContactUsLinkComponent
Human recombinant nectin-2 (from HEK293)
Human recombinant nectin-2 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant nectin-2 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7,5, Normally trehalose is added as protectant before Lyophilisation,; Measured by its binding ability in a functional ELISA, Immobilised Human PVRIG, His Tag (Cat, No, PVG-H52H4) at 2 μg/ml (100 μL/well) can bind Human Nectin-2, Fc Tag (Cat, No, PV2-H5253) with a linear range of 0,5-8 ng/mL (QC tested),; Measured by its binding ability in a functional ELISA, Immobilised Human Nectin-2, Fc Tag (Cat, No, PV2-H5253) at 10 μg/ml (100 μL/well) can bind Biotinylated Human TIGIT, Fc Tag, Avi Tag with a linear range of 0,156-2,5 μg/ml (Routinely tested),; Loaded Human Nectin-2, Fc Tag (Cat, No, PV2-H5253) on Protein A Biosensor, can bind Human PVRIG, His Tag (Cat, No, PVG-H52H4) with an affinity constant of 0,456 μM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    Q92692-2
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    Nectin-2|PVRL2|CD112
  • Protein/peptide name:
    Nectin-2
  • Purity:
    >95%
  • Molecular weight:
    62,2 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5, Normally trehalose is added as protectant before Lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together