Order Entry
Austria
ContactUsLinkComponent
Human recombinant LILRB5 (from HEK293)
Human recombinant LILRB5 (from HEK293)
Supplier:  ACROBIOSYSTEMS
Certificates
Human recombinant LILRB5 (from HEK293)
Supplier:  ACROBIOSYSTEMS

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Biological activity:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before Lyophilisation.
  • Gene ID:
    O75023-1
  • Carrier-free:
    Y
  • Protease-free:
    Y
  • Animal-free:
    Y
  • Protein synonyms:
    LIR-8|CD85c|LILRB5
  • Protein/peptide name:
    LILRB5
  • Purity:
    >95%
  • Molecular weight:
    75,8 kDa
  • Endotoxin level:
    Low
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5. Normally trehalose is added as protectant before Lyophilisation.
  • Nuclease-free:
    Y
  • Shipping temperature:
    Powder,RT

Specifications

Frequently Bought Together