Human recombinant LAIR-1 (from HEK293)
Supplier: ACROBIOSYSTEMS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Less than 1,0 EU per μg by the LAL method.
- Biological activity:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5, Normally trehalose is added as protectant before Lyophilisation,; Measured by its binding ability in a functional ELISA, Immobilised Human Collagen I protein at 2 μg/ml (100 μl/well) can bind Human LAIR-1, Fc Tag (Cat, No, LA1-H5252) with a linear range of 20-156 ng/ml (QC tested).
- Gene ID:Q6GTX8-1
- Carrier-free:Y
- Protease-free:Y
- Animal-free:Y
- Protein synonyms:LAIR1|CD305
- Protein/peptide name:LAIR-1
- Purity:>95%
- Molecular weight:42,1 kDa
- Endotoxin level:Low
- Formulation:Lyophilised from 0,22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH 7,5, Normally trehalose is added as protectant before Lyophilisation.
- Nuclease-free:Y
- Shipping temperature:Powder,RT
Specifications
Frequently Bought Together