Order Entry
Austria
ContactUsLinkComponent
 
Sodium hydroxide solution 0.1 N (0.1 M), in single-use containers, HyClone™ for bioprocessing
 
Sodium hydroxide solution 0.1 N (0.1 M), in single-use containers, HyClone™ for bioprocessing

 

  • Storage:
    15° to 30°C
  • Certifications/Compliance:
    Manufactured in a ISO9001 and ISO13485 certified facility
  • Shelf life:
    12 Months
  • pH:
    For Information Only
  • For use with:
    Chromatography resins, IC systems, ÄKTA hardware systems
  • Appearance:
    Clear, colorless solution
  • Sterility:
    No Growth

 

Frequently Bought Together