Orders LinkContactUsLinkComponent
CF® Dye Dextran 250000 MW, anionic and fixable
CF® Dye Dextran 250000 MW, anionic and fixable
Supplier:   Biotium
CF® Dye Dextran 250000 MW, anionic and fixable
Supplier:  Biotium
Certificates
CF® Dye Dextran 250000 MW, anionic and fixable
CF® Dye Dextran 250000 MW, anionic and fixable
4 Options Available Below
Product Details & Documents
CF® dye labeled dextrans could be used as a fluorescent fluid-phase markers to study cell permeability, endocytosis, or mechanisms of biomolecular delivery. The dextran is 250,000 MW, and contains a formaldehyde-fixable free-amine group.
  • Water-soluble fluid phase markers for tracking bulk endocytosis and cell permeability
  • Available with molecular weights of 3.5 kD, 10 kD, 40 kD, 70kD, 150 kD, or 250 kD
  • Choose from a selection of bright & photostable CF® dyes
Dextrans are water soluble branched-chain polysaccharides. Fluorescently labeled dextrans are used as markers for trafficking of fluid phase endocytic cargo to lysosomes. They are also used as tracers for epithelial and endothelial permeability. Labeled dextrans may also be used as microinjected tracers for neuronal morphology. CF® Dye Dextrans are available with a variety of dye colors and molecular weights ranging from 3,500 to 250,000.
These conjugates are available with Biotium's next-generation CF® dyes, a series of highly water-soluble fluorescent dyes spanning the visible and near-infrared spectrum for labeling biomolecules such as proteins and nucleic acids. Developed in house by Biotium's industry-leading chemists, CF® dyes offer superior brightness, photostability, and signal-to-noise when compared to other commercially available fluorescent dyes.
Product Family Options
Product Information
  • Ex/Em
    Pk
    Availability
    Price
  • 80134.
    CF488A-Dextran 250,000 MW, anionic and fixable
    CF488A-Dextran 250,000 MW, anionic and fixable
    490/515 nm
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 169,00 €
    169,00 €
    1 * 1 mg
       169,00 €
  • 80135.
    CF680-Dextran 250,000 MW, anionic and fixable
    CF680-Dextran 250,000 MW, anionic and fixable
    681/698 nm
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 197,00 €
    197,00 €
    1 * 1 mg
       197,00 €
  • 80136.
    CF750-Dextran 250,000 MW, anionic and fixable
    CF750-Dextran 250,000 MW, anionic and fixable
    755/777 nm
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 197,00 €
    197,00 €
    1 * 1 mg
       197,00 €
  • 80125.
    CF770-Dextran 250,000 MW, anionic and fixable
    CF770-Dextran 250,000 MW, anionic and fixable
    770/797 nm
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 197,00 €
    197,00 €
    1 * 1 mg
       197,00 €