VWR®, PETG Erlenmeyer Shaker Flask
VWR® crystal-clear PETG Erlenmeyer shaker flasks are robust and break resistant, increasing personal safety. They are ideal for shaker culture applications and storage.
- Temperature range: −60 to +60 °C
- Unlike glass, flasks will not break when dropped
- Moulded-in graduations for accuracy
- Vent cap option for continuous gas exchange while ensuring sterility and preventing leakage
- Non cytotoxic, DNase-/RNase-free and non pyrogenic
- Sterilised by irradiation SAL 10⁻⁶ (ISO 11137)
- Shelf life: 3 years after the month of production
Manufactured in a Class 100,000 room environment under ISO 13485 and ISO 9001 quality management systems.
: Available with plug seal cap or vent cap.
: Individually packed.
- Catalog No:
- 214-0442
- 214-0440
- 214-0445
- 214-0455
- 214-0454
- 214-0444
- 214-0443
- 214-0441
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Frequently Bought Together