Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Fluoro-N-methyl-4-nitrobenzamide
3-Fluoro-N-methyl-4-nitrobenzamide 95%
3-Fluoro-N-methyl-4-nitrobenzamide 95%
APOSPC907963-1G
:
Apollo Scientific
:
3-Fluoro-N-methyl-4-nitrobenzamide 95%
APOSPC907963-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
198.151 g/mol
:
Ambient
:
658700-20-0
:
C₈H₇FN₂O₃
:
MFCD14583038