N-(Oct-1-yl)-perfluoro-2,5-dimethyl-3,6-dioxanonanamide
Catalog # APOSPC6939-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:607.303 g/mol
Storage Temperature:Ambient
CAS Number:340037-41-4
Formula:C₁₇H₁₈F₁₇NO₃
MDL Number:MFCD00310125