1,2,4,5-Tetrafluoro-3,6-bis(2-phenylethynyl)benzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1408279-25-3
:
Formula:C₂₂H₁₀F₄
MDL Number:MFCD01596892
:350.308 g/mol
Storage Temperature:Ambient
Frequently Bought Together