Methyl 5-bromo-6-fluoronicotinate 95+%
Catalog # APOSPC535044-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:234.023 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:405939-62-0
Formula:C₇H₅BrFNO₂
MDL Number:MFCD14697990